Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028315.2 | NP_082591.2 | 15 | 2048 | ||
GenBank mRNA | AK032091.1 | - | 10 | 1138 | ||
AK038313.1 | - | 15 | 1918 | |||
BC027357.1 | - | 14 | 1933 | 4952415 |