Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146024.1 | NP_001139496.1 | 5 | 2020 | ||
NM_028316.3 | NP_082592.2 | 4 | 1900 | |||
GenBank mRNA | AK089886.1 | - | 4 | 1869 | ||
AK220307.1 | - | 1338 |