Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skida1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028317.2 | NP_082593.2 | 1 | 2525 | ||
GenBank mRNA | AK012843.1 | - | 1 | 72 | ||
AK141728.1 | - | 1 | 1078 | |||
BC050860.1 | - | 2 | 2264 | 6331376 |