Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144802.4 | NP_659051.3 | 8, 9 | 1509 | ||
GenBank mRNA | AK012866.1 | - | 8, 9 | 1225 | ||
AK146879.1 | - | 8, 9 | 1471 | |||
BC004763.1 | - | 5, 6 | 482 | |||
BC012849.1 | - | 8, 9 | 1455 | 3587716 |