Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef10l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112722.1 | NP_001106193.1 | 23 | 2666 | ||
NM_001112723.1 | NP_001106194.1 | 22 | 2624 | |||
NM_001290803.1 | NP_001277732.1 | 24 | 2765 | |||
NM_172415.3 | NP_766003.3 | 23 | 2741 | |||
GenBank mRNA | AK028570.1 | - | 17 | 2082 | ||
AK028648.1 | - | 23 | 2649 | |||
AK129408.1 | - | 22 | 2624 | |||
BC021843.1 | - | 9 | 911 | |||
BC034061.1 | - | 17 | 1902 | |||
BC059220.1 | - | 23 | 2729 | 6417641 | ||
CA751244.1 | - | 3 | 319 | |||
CF531696.1 | - | 2 | 163 |