Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pot1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028370.1 | NP_082646.1 | 8 | 1033 | ||
GenBank mRNA | AK045286.1 | - | 8 | 984 | ||
AK050727.1 | - | 8 | 1032 | |||
AK165040.1 | - | 8 | 1010 |