Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293228.1 | NP_001280157.1 | 3 | 1685 | ||
NM_001293231.1 | NP_001280160.1 | 2 | 468 | |||
NM_001293233.1 | NP_001280162.1 | 3 | 1375 | |||
NM_005378.5 | NP_005369.2 | 3 | 1375 | |||
XM_017004168.1 | XP_016859657.1 | 2 | 1206 | |||
GenBank mRNA | AK298225.1 | - | 4 | 668 | ||
AK299495.1 | - | 4 | 1003 | |||
BC002712.2 | - | 2 | 1093 | 3627330 | ||
BT007384.1 | - | 2 | 952 |