Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278415.1 | NP_001265344.1 | 9 | 885 | ||
NM_028388.3 | NP_082664.1 | 8 | 788 | |||
GenBank mRNA | AK013511.1 | - | 8 | 739 | ||
AK078351.1 | - | 8 | 732 | |||
AK088193.1 | - | 5 | 527 | |||
AK146998.1 | - | 8 | 722 | |||
AK150404.1 | - | 9 | 822 | |||
AK151729.1 | - | 9 | 822 | |||
AK159460.1 | - | 8 | 705 | |||
AK166539.1 | - | 8 | 749 | |||
AK169143.1 | - | 8 | 759 | |||
BC030946.1 | - | 8 | 717 | 5003524 |