Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293784.1 | NP_001280713.1 | 3 | 394 | ||
NM_175188.4 | NP_780397.2 | 3 | 490 | |||
GenBank mRNA | AK013582.1 | - | 3 | 396 | ||
AK086052.1 | - | 3 | 353 | |||
AK160684.1 | - | 3 | 323 | |||
BB662698.1 | - | 3 | 394 | |||
BC066008.1 | - | 3 | 490 | 5703753 |