Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028399.1 | NP_082675.1 | 2 | 251 | ||
GenBank mRNA | AK013634.1 | - | 2 | 198 | ||
AK036772.1 | - | 199 | ||||
AK049863.1 | - | 2 | 196 | |||
AK152399.1 | - | 2 | 199 | |||
AK154439.1 | - | 2 | 209 | |||
AK156231.1 | - | 220 | ||||
AK156621.1 | - | 2 | 207 | |||
BC054122.1 | - | 2 | 251 | 6509024 | ||
JX965957.1 | - | 2 | 187 | |||
JX965958.1 | - | 2 | 187 |