Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182993.2 | NP_892038.2 | 7 | 903 | ||
GenBank mRNA | AK044409.1 | - | 1 | 1916 | ||
AK159064.1 | - | 7 | 876 | |||
BC028938.1 | - | 5 | 342 | |||
BC054462.1 | - | 5 | 685 | 6494471 |