Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccser2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027045.1 | NP_081321.1 | 11 | 3243 | ||
NM_028407.3 | NP_082683.2 | 8 | 1330 | |||
GenBank mRNA | AK005931.1 | - | 8 | 1330 | ||
AK147383.1 | - | 11 | 3237 | |||
BC111465.1 | - | 9 | 1539 | |||
CV556741.1 | - | 1 | 187 |