Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145221.2 | NP_660256.1 | 2 | 164 | ||
GenBank mRNA | AK013715.1 | - | 2 | 161 | ||
AK030445.1 | - | 2 | 44 | |||
AK033566.1 | - | 2 | 135 | |||
AK045438.1 | - | 2 | 100 | |||
AK049307.1 | - | 2 | 151 | |||
AK088080.1 | - | 2 | 115 | |||
AK146356.1 | - | 2 | 135 | |||
AY113705.1 | - | 2 | 129 | |||
BC063751.1 | - | 2 | 176 | 6511401 | ||
BU514763.1 | - | 2 | 175 |