Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Appl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145221.2 | NP_660256.1 | 9 | 772 | ||
GenBank mRNA | AK030445.1 | - | 9 | 652 | ||
AK033566.1 | - | 9 | 743 | |||
AK045438.1 | - | 9 | 708 | |||
AK049307.1 | - | 9 | 759 | |||
AK146356.1 | - | 9 | 743 | |||
AY113705.1 | - | 9 | 737 | |||
BC019708.1 | - | 1 | 50 | |||
BC063751.1 | - | 9 | 784 | 6511401 | ||
BU514763.1 | - | 9 | 783 |