Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271531.1 | NP_001258460.1 | 5 | 830 | ||
NM_001271532.1 | NP_001258461.1 | 4 | 697 | |||
NM_133748.2 | NP_598509.1 | 5 | 1073 | |||
NM_178082.3 | NP_835183.1 | 5 | 837 | |||
GenBank mRNA | AF527631.1 | - | 5 | 807 | ||
AK040703.1 | - | 5 | 828 | |||
AK050394.1 | - | 5 | 831 | |||
AK152076.1 | - | 5 | 803 | |||
AK154543.1 | - | 5 | 836 | |||
AK161256.1 | - | 5 | 841 | |||
AK165634.1 | - | 5 | 842 | |||
AK191083.1 | - | 3 | 324 | |||
AK192853.1 | - | 3 | 282 | |||
BC015288.1 | - | 5 | 787 | 4240711 | ||
BC023067.1 | - | 5 | 811 | 5357599 | ||
BC023874.1 | - | 4 | 729 | 5355039 | ||
BC024411.1 | - | 5 | 825 | 4971297 |