Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271531.1 | NP_001258460.1 | 4 | 729 | ||
NM_001271532.1 | NP_001258461.1 | 3 | 596 | |||
NM_133748.2 | NP_598509.1 | 4 | 972 | |||
NM_178082.3 | NP_835183.1 | 4 | 736 | |||
GenBank mRNA | AF527631.1 | - | 4 | 706 | ||
AK040703.1 | - | 4 | 727 | |||
AK050394.1 | - | 4 | 730 | |||
AK152076.1 | - | 4 | 702 | |||
AK154543.1 | - | 4 | 735 | |||
AK161256.1 | - | 4 | 740 | |||
AK165634.1 | - | 4 | 741 | |||
AK190310.1 | - | 2 | 207 | |||
AK191083.1 | - | 2 | 223 | |||
AK192853.1 | - | 2 | 181 | |||
BC015288.1 | - | 4 | 686 | 4240711 | ||
BC023067.1 | - | 4 | 710 | 5357599 | ||
BC023874.1 | - | 3 | 628 | 5355039 | ||
BC024411.1 | - | 4 | 724 | 4971297 |