Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r16a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033371.2 | NP_203535.1 | 4 | 1148 | ||
GenBank mRNA | AK154865.1 | - | 4 | 1133 | ||
AK171452.1 | - | 4 | 1133 | |||
AK220248.1 | - | 4 | 1148 | |||
AY010723.1 | - | 3 | 343 | |||
BC033318.1 | - | 3 | 708 | |||
BC044775.1 | - | 3 | 830 | |||
BC049958.1 | - | 3 | 830 | |||
BC057450.1 | - | 3 | 765 | 3966334 | ||
BC096620.1 | - | 4 | 1148 | 6390076 |