Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmpcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028431.2 | NP_082707.1 | 9 | 1160 | ||
GenBank mRNA | AK013995.1 | - | 9 | 1161 | ||
AK169417.1 | - | 9 | 1117 | |||
BC038321.1 | - | 8 | 1031 | |||
BC051216.1 | - | 8 | 967 | |||
BC060662.1 | - | 8 | 3672 | |||
BC065095.1 | - | 8 | 3672 | |||
BC087943.1 | - | 9 | 1172 |