Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028876.2 | NP_083152.1 | 4 | 691 | ||
GenBank mRNA | AK014490.1 | - | 4 | 692 | ||
AK033463.1 | - | 4 | 619 | |||
AK151650.1 | - | 4 | 657 | |||
AK159813.1 | - | 3 | 1779 | |||
AK170036.1 | - | 4 | 651 | |||
BC020076.1 | - | 4 | 653 | 3985782 | ||
BC132023.1 | - | 4 | 653 | 40130300 | ||
BC138226.1 | - | 4 | 653 | 8861245 |