Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028454.2 | NP_082730.2 | 4, 5 | 637 | ||
GenBank mRNA | AF480070.1 | - | 4, 5 | 511 | ||
AK014166.1 | - | 4, 5 | 618 | |||
AK037325.1 | - | 3, 4 | 441 | |||
AK039028.1 | - | 4, 5 | 567 | |||
AK040016.1 | - | 4, 5 | 513 | |||
AK043687.1 | - | 3 | 904 | |||
AK134512.1 | - | 1 | 1422 | |||
AK155475.1 | - | 4, 5 | 594 | |||
BC014769.1 | - | 4, 5 | 629 | 4239159 |