Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028454.2 | NP_082730.2 | 7 | 885 | ||
GenBank mRNA | AF480070.1 | - | 7 | 759 | ||
AK014166.1 | - | 7 | 866 | |||
AK155475.1 | - | 7 | 842 | |||
BC014769.1 | - | 7 | 877 | 4239159 |