Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164627.1 | NP_001158099.1 | 13 | 1300 | ||
NM_001164628.1 | NP_001158100.1 | 12 | 1292 | |||
NM_001205334.1 | NP_001192263.1 | 13 | 1320 | |||
NM_028455.4 | NP_082731.2 | 12 | 1312 | |||
GenBank mRNA | AK014171.1 | - | 12 | 1263 | ||
AK086816.1 | - | 12 | 1266 | |||
AK088242.1 | - | 12 | 1268 | |||
BC005563.1 | - | 11 | 1255 | 3965919 | ||
BC010306.1 | - | 12 | 1282 |