Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164627.1 | NP_001158099.1 | 11 | 921 | ||
NM_001164628.1 | NP_001158100.1 | 10 | 913 | |||
NM_001205334.1 | NP_001192263.1 | 11 | 941 | |||
NM_028455.4 | NP_082731.2 | 10 | 933 | |||
GenBank mRNA | AK014171.1 | - | 10 | 884 | ||
AK086816.1 | - | 10 | 887 | |||
AK088242.1 | - | 10 | 889 | |||
BC005563.1 | - | 9 | 876 | 3965919 | ||
BC010306.1 | - | 10 | 903 |