Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rwdd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025637.3 | NP_079913.2 | 4 | 655 | ||
NM_028456.3 | NP_082732.2 | 2 | 439 | |||
GenBank mRNA | AF439556.1 | - | 2 | 438 | ||
AF439557.1 | - | 3 | 645 | |||
AK014130.1 | - | 2 | 377 | |||
AK134288.1 | - | 2 | 430 | |||
AK151329.1 | - | 3 | 670 | |||
AK151467.1 | - | 3 | 670 | |||
AK153551.1 | - | 3 | 670 | |||
BC128287.1 | - | 2 | 438 | 40102432 | ||
BC128288.1 | - | 4 | 654 | |||
BC139340.1 | - | 2 | 438 | 8862362 | ||
BC139342.1 | - | 2 | 438 | 8862364 |