Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032413.1 | NP_001027585.1 | 10 | 1154 | ||
NM_001032414.1 | NP_001027586.1 | 9 | 1084 | |||
NM_001289600.1 | NP_001276529.1 | 9 | 1056 | |||
NM_001289601.1 | NP_001276530.1 | 11 | 1297 | |||
NM_028460.2 | NP_082736.1 | 11 | 1288 | |||
NM_152799.2 | NP_690012.2 | 11 | 1253 | |||
GenBank mRNA | AF440279.1 | - | 11 | 1278 | ||
AF444274.1 | - | 9 | 1078 | |||
AF461685.1 | - | 8 | 1011 | |||
AK042308.1 | - | 9 | 1470 | |||
AK053551.1 | - | 10 | 1145 | |||
AK141082.1 | - | 1051 | ||||
AK161049.1 | - | 11 | 1294 | |||
BC042490.1 | - | 10 | 1103 | 4953584 |