Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pear1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032413.1 | NP_001027585.1 | 3, 4 | 316 | ||
NM_001032414.1 | NP_001027586.1 | 2, 3 | 246 | |||
NM_001289600.1 | NP_001276529.1 | 2, 3 | 246 | |||
NM_001289601.1 | NP_001276530.1 | 4, 5 | 450 | |||
NM_028460.2 | NP_082736.1 | 4, 5 | 450 | |||
NM_152799.2 | NP_690012.2 | 4, 5 | 415 | |||
GenBank mRNA | AF440279.1 | - | 4, 5 | 440 | ||
AF444274.1 | - | 2, 3 | 240 | |||
AF461685.1 | - | 1, 2 | 173 | |||
AK014179.1 | - | 2, 3 | 250 | |||
AK042308.1 | - | 3, 4 | 307 | |||
AK053551.1 | - | 3, 4 | 307 | |||
AK141082.1 | - | 241 | ||||
AK161049.1 | - | 4, 5 | 447 | |||
BC042490.1 | - | 3, 4 | 265 | 4953584 |