Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 3110043O21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081343.1 | NP_001074812.1 | 3 | 609 | ||
GenBank mRNA | AK014175.1 | - | 2 | 61 | ||
AK154817.1 | - | 3 | 583 | |||
AK167354.1 | - | 3 | 540 | |||
BC026738.1 | - | 1 | 14 | 4020832 | ||
BC076612.1 | - | 3 | 711 | 30638238 |