Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam118a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133750.4 | NP_598511.1 | 3 | 472 | ||
GenBank mRNA | AK078807.1 | - | 1 | 210 | ||
AK082151.1 | - | 3 | 410 | |||
AK143690.1 | - | 3 | 445 | |||
BC016425.1 | - | 3 | 330 | 4487028 | ||
BC058794.1 | - | 3 | 533 | 6486049 | ||
BC066174.1 | - | 3 | 388 | 30109713 |