Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000257.3 | NP_000248.2 | 10 | 976 | ||
XM_017021340.1 | XP_016876829.1 | 9 | 911 | |||
GenBank mRNA | AK307590.1 | - | 10 | 950 | ||
BC112171.1 | - | 9 | 900 | 8327639 | ||
BC112173.1 | - | 9 | 900 | 8327641 | ||
EF560725.1 | - | 8 | 843 | |||
EU747717.1 | - | 10 | 951 | |||
M58018.1 | - | 10 | 931 |