Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700034I23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006596.1 | - | 1 | 200 | ||
AK077003.1 | - | 1 | 158 | |||
BC117739.1 | - | 1 | 166 | 40090015 |
Gene Symbol: Gm10731 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077188.1 | - | 1 | 151 | ||
BY081654.1 | - | 154 |