Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270435.1 | NP_001257364.1 | 14 | 2155 | ||
NM_001270446.1 | NP_001257375.1 | 9 | 1411 | |||
NM_028506.1 | NP_082782.1 | 10 | 1418 | |||
NM_029286.2 | NP_083562.1 | 14 | 2170 | |||
GenBank mRNA | AK049010.1 | - | 9 | 1311 | ||
AK078362.1 | - | 14 | 2097 | |||
AK163185.1 | - | 14 | 2096 | |||
AK224940.1 | - | 10 | 1347 | |||
AK224961.1 | - | 9 | 1410 | |||
BC114436.1 | - | 14 | 2097 | 40059415 |