Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itpripl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163527.1 | NP_001156999.1 | 2 | 1035 | ||
NM_001163528.1 | NP_001157000.1 | 2 | 859 | |||
GenBank mRNA | AK006667.1 | - | 2 | 854 | ||
AK028913.1 | - | 3 | 775 | |||
BC130218.1 | - | 2 | 884 | 40141227 |