Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152801.2 | NP_690014.2 | 16 | 1999 | ||
GenBank mRNA | AF393831.1 | - | 16 | 1999 | ||
AK040468.1 | - | 16 | 1821 |