Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phospho2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028521.2 | NP_082797.1 | 4 | 716 | ||
GenBank mRNA | AK006724.1 | - | 4 | 717 | ||
AK132362.1 | - | 2 | 1844 | |||
AK159551.1 | - | 4 | 763 | |||
BC025612.1 | - | 4 | 752 | 5257820 | ||
BC031523.1 | - | 2 | 387 | 4009719 |