Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040113.1 | NP_001035202.1 | 8, 9 | 916 | ||
NM_001040114.1 | NP_001035203.1 | 8, 9 | 916 | |||
NM_002474.2 | NP_002465.1 | 7, 8 | 895 | |||
NM_022844.2 | NP_074035.1 | 7, 8 | 895 | |||
XM_011522502.2 | XP_011520804.1 | 7, 8 | 897 | |||
XM_017023250.1 | XP_016878739.1 | 8, 9 | 880 | |||
GenBank mRNA | AB020673.2 | - | 7, 8 | 863 | ||
AK125033.1 | - | 2, 3 | 1210 | |||
AK308389.1 | - | 7, 8 | 892 | |||
AY520816.1 | - | 8, 9 | 897 | |||
AY520817.1 | - | 8, 9 | 897 | |||
BC101677.1 | - | 7, 8 | 859 | 8069183 | ||
BC104906.1 | - | 7, 8 | 859 | 8143909 | ||
BC143364.1 | - | 8, 9 | 880 | 9051869 | ||
EU489063.1 | - | 8, 9 | 916 |
Gene Symbol: MYH9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KU159427.1 | - | 989 |