Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam177a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028527.1 | NP_082803.1 | 2 | 268 | ||
GenBank mRNA | AK006713.1 | - | 101 | |||
AK045530.1 | - | 328 | ||||
AK159923.1 | - | 306 | ||||
BC048158.1 | - | 268 | 5698044 | |||
BC049669.1 | - | 327 | 6535412 |
Gene Symbol: 1700047I17Rik2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100116.1 | NP_001093586.1 | 268 |