Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290783.1 | NP_001277712.1 | 4 | 967 | ||
NM_026137.5 | NP_080413.2 | 4 | 1001 | |||
NM_028542.1 | NP_082818.1 | 4 | 1253 | |||
GenBank mRNA | AF220146.1 | - | 3 | 523 | ||
AF353243.1 | - | 4 | 647 | |||
AF513713.1 | - | 4 | 470 | |||
AK006837.1 | - | 3 | 489 | |||
AK017549.1 | - | 4 | 583 | |||
BC037502.1 | - | 4 | 618 | |||
BC057366.1 | - | 4 | 967 | |||
BC080853.1 | - | 4 | 916 | |||
BC108332.1 | - | 4 | 757 | 6493226 | ||
BC120553.1 | - | 4 | 504 | 40129476 | ||
BC125287.1 | - | 4 | 503 | 40129802 |