Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700067K01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183097.2 | NP_898920.2 | 4 | 555 | ||
GenBank mRNA | AK006917.1 | - | 4 | 824 | ||
BC147276.1 | - | 4 | 561 | 9056316 | ||
BC147277.1 | - | 4 | 561 | 9056320 | ||
BC172025.1 | - | 4 | 558 | 9054311 | ||
BC172026.1 | - | 4 | 558 |