Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pebp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028526.4 | NP_082802.1 | 3 | 449 | ||
NM_028560.4 | NP_082836.2 | 3 | 449 | |||
GenBank mRNA | AK006750.1 | - | 3 | 450 | ||
AK006964.1 | - | 3 | 440 | |||
BC048621.1 | - | 3 | 427 | 6742909 |