Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183099.2 | NP_898922.2 | 1 | 410 | ||
GenBank mRNA | AK007032.1 | - | 1 | 412 | ||
BC114543.1 | - | 1 | 214 |