Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002841.1 | NP_001002841.1 | 8 | 772 | ||
NM_002476.2 | NP_002467.1 | 7 | 719 | |||
XM_005257391.4 | XP_005257448.1 | 8 | 699 | |||
XM_011524839.2 | XP_011523141.2 | 8 | 859 | |||
XM_017024683.1 | XP_016880172.1 | 8 | 846 | |||
GenBank mRNA | AF116676.2 | - | 1071 | |||
BC030228.1 | - | 7 | 708 | 5226518 | ||
M20641.1 | - | 4 | 335 | |||
M24121.1 | - | 7 | 706 | |||
M36172.1 | - | 7 | 700 | |||
X13955.1 | - | 7 | 700 | |||
X52005.1 | - | 725 |