Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700001O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198000.3 | NP_932117.2 | 2 | 620 | ||
GenBank mRNA | AK005618.1 | - | 2 | 131 | ||
AK007200.1 | - | 109 | ||||
AK019754.1 | - | 2 | 97 | |||
AK076928.1 | - | 2 | 121 | |||
BC023044.1 | - | 2 | 590 | 5355351 |