Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026865.2 | NP_081141.1 | 2 | 751 | ||
GenBank mRNA | AK004415.1 | - | 2 | 69 | ||
AK007198.1 | - | 2 | 750 | |||
BC057970.1 | - | 2 | 57 | |||
BC141198.1 | - | 2 | 170 | 9055810 |