Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cybrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028593.2 | NP_082869.2 | 4 | 676 | ||
GenBank mRNA | AF354666.1 | - | 4 | 618 | ||
AK008849.1 | - | 4 | 622 | |||
AK029112.1 | - | 4 | 679 | |||
BC027665.1 | - | 2 | 172 | |||
BC147289.1 | - | 2 | 164 | |||
BC147290.1 | - | 2 | 164 |