Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002477.1 | NP_002468.1 | 2 | 176 | ||
XM_006713886.2 | XP_006713949.1 | 2 | 553 | |||
XM_017008245.1 | XP_016863734.1 | 3 | 1854 | |||
XM_017008246.1 | XP_016863735.1 | 4 | 1723 | |||
XM_017008247.1 | XP_016863736.1 | 2 | 1313 | |||
XM_017008248.1 | XP_016863737.1 | 3 | 336 | |||
GenBank mRNA | AK123974.1 | - | 3 | 2472 | ||
AK125529.1 | - | 3 | 1156 | |||
BC040050.1 | - | 2 | 296 | 6014371 | ||
L03785.1 | - | 2 | 176 |