Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166376.1 | NP_001159848.1 | 3 | 426 | ||
NM_028595.4 | NP_082871.2 | 3 | 426 | |||
GenBank mRNA | AF237910.1 | - | 3 | 384 | ||
AK008652.1 | - | 3 | 400 | |||
AK075852.1 | - | 3 | 354 | |||
AK150397.1 | - | 3 | 323 | |||
AK152258.1 | - | 3 | 323 | |||
BC062247.1 | - | 3 | 369 | 1550331 |