Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047604.2 | NP_001041069.1 | 18 | 2490 | ||
NM_001290669.1 | NP_001277598.1 | 18 | 2490 | |||
GenBank mRNA | AK132240.1 | - | 4 | 409 | ||
AK164723.1 | - | 8 | 1117 | |||
BC151199.1 | - | 18 | 2465 | |||
BC172115.1 | - | 18 | 2464 | |||
DQ011164.1 | - | 18 | 2490 |