Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc21b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047604.2 | NP_001041069.1 | 14 | 1772 | ||
NM_001290669.1 | NP_001277598.1 | 14 | 1772 | |||
GenBank mRNA | AK147810.1 | - | 7 | 855 | ||
AK164723.1 | - | 4 | 399 | |||
BC151199.1 | - | 14 | 1747 | |||
BC172115.1 | - | 14 | 1746 | |||
DQ011164.1 | - | 14 | 1772 |