Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dppa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139218.1 | NP_631964.1 | 2 | 214 | ||
GenBank mRNA | AB072734.1 | - | 2 | 115 | ||
AF490347.1 | - | 2 | 219 | |||
AK010723.1 | - | 2 | 152 | |||
AK135769.1 | - | 2 | 219 | |||
AK136263.1 | - | 2 | 219 | |||
AY082485.1 | - | 2 | 214 | |||
AY134859.1 | - | 2 | 226 | |||
BC099433.1 | - | 2 | 192 | 30933569 | ||
BC100331.1 | - | 2 | 189 | 30934920 | ||
BC107340.1 | - | 2 | 146 | 40055218 | ||
BC107341.1 | - | 2 | 146 | 40055220 |