Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1110017D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048005.1 | NP_001041470.1 | 6 | 904 | ||
NM_001253777.1 | NP_001240706.1 | 6 | 909 | |||
NM_001253778.1 | NP_001240707.1 | 3 | 516 | |||
NM_001253780.1 | NP_001240709.1 | 2 | 281 | |||
NM_001253788.1 | NP_001240717.1 | 5 | 674 | |||
NM_028624.1 | NP_082900.1 | 3 | 511 | |||
GenBank mRNA | AB231294.1 | - | 6 | 908 | ||
AK003742.1 | - | 3 | 502 | |||
BC049629.1 | - | 3 | 442 | |||
CA463601.1 | - | 3 | 514 | |||
DQ873295.1 | - | 3 | 373 | |||
DQ873296.1 | - | 3 | 378 |